Anti-Glucose Transporter GLUT3+GLUT14 Rabbit Monoclonal Antibody [clone: EPR28481-54]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB315169-1MG AB315169-100UG
ABCAAB315169-1MGEA 9160 EUR
ABCAAB315169-1MG ABCAAB315169-100UG
Anti-Glucose Transporter GLUT3+GLUT14 Rabbit Monoclonal Antibody [clone: EPR28481-54]
Antilichamen
Rabbit monoclonal [EPR28481-54] to Glucose Transporter GLUT3 + GLUT14 - BSA and Azide free.

Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Glucose Transporter GLUT3+GLUT14
Clonality: Monoclonal
Clone: EPR28481-54
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR