Anti-CaMKII beta Rabbit Monoclonal Antibody [clone: EPR28076-54]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB314897-20UL AB314897-100UL AB314897-1ML
ABCAAB314897-20ULEA 244 EUR
ABCAAB314897-20UL ABCAAB314897-100UL ABCAAB314897-1ML
Anti-CaMKII beta Rabbit Monoclonal Antibody [clone: EPR28076-54]
Antilichamen
Rabbit monoclonal [EPR28076-54] to CaMKII beta.
Opgelet: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CaMKII beta
Clonality: Monoclonal
Clone: EPR28076-54
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat