Anti-MYL9+MYL12A+MYL12B (phospho S20) Rabbit Monoclonal Antibody [clone: EPR27203-65]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AB316750-20UL AB316750-100UL AB316750-1ML
ABCAAB316750-20ULEA 244 EUR
ABCAAB316750-20UL ABCAAB316750-100UL ABCAAB316750-1ML
Anti-MYL9+MYL12A+MYL12B (phospho S20) Rabbit Monoclonal Antibody [clone: EPR27203-65]
Antilichamen
Rabbit monoclonal [EPR27203-65] to MYL9 (phospho S20) + MYL12A (phospho S19) + MYL12B (phospho S20).

Opgelet: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MYL9+MYL12A+MYL12B
Clonality: Monoclonal
Clone: EPR27203-65
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR