rubiflex S NB35SF Chemical Protection Gloves, 60209 Series
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6020908 6020909 6020910 6020911
UVEX6020908EA 261 EUR
UVEX6020908 UVEX6020909 UVEX6020910 UVEX6020911
rubiflex S NB35SF Chemical Protection Gloves, 60209 Series
Handschoenen
This chemical protection glove features a double cotton-interlock lining. This means that the rubiflex SNB35SF is suitable for a wide range of applications that require chemical protection in addition to increased mechanical protection to heat and cold.
  • NBR chemical protection glove with double cotton liner on the palm
  • Suitable for working with food
  • Cotton lining ensures excellent moisture absorption
  • AQL: 1,5
The uvex rubiflex S chemical protection gloves are characterised by excellent wearer comfort, the good mechanical abrasion resistance of the coating and good resistance to many chemicals, acids, alkalis, mineral oils and solvents. Their ergonomic fit means that they have a good tactile feel and are very flexible. The rubiflex S NB35SF is particularly suited to damp, cold, and dry working conditions, but can also be used when working with contact heat up to 250 °C. Free from harmful substances in line with OEKO-TEX® standard 100.