PointGuard® Ultra 9032 Safety Gloves, 60638 Series
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6063810 6063807 6063808 6063809
UVEX6063810EA 0 EUR
UVEX6063810 UVEX6063807 UVEX6063808 UVEX6063809
PointGuard® Ultra 9032 Safety Gloves, 60638 Series
Handschoenen
The hi-vis PointGuard® Ultra 9032 provides industry-leading cut and needlestick protection, complete with a hi-vis polyester shell and a sandy nitrile 3/4 knuckle coating.
  • High needlestick and cut resistance
  • Sandy nitrile 3/4 knuckle palm coating over polyester shell
  • Seamless construction delivers high comfort and dexterity
Offering superior grip and abrasion resistance in wet conditions, these gloves provide the highest needlestick and cut resistance available due to the layering of SuperFabric®* brand material on the palm. Three layers of SuperFabric® material protect against hazards through a configuration of tiny guard plates layered onto performance fabrics. The 15-gauge hi-vis polyester shell enhances visibility and worker compliance. Knit wrist helps prevent dirt and debris from entering the glove.