HexArmor® SharpsMaster II® 9014 Safety Gloves, 60981 Series
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6098106 6098107 6098108 6098109 6098110
UVEX6098106EA 0 EUR
UVEX6098106 UVEX6098107 UVEX6098108 UVEX6098109 UVEX6098110
HexArmor® SharpsMaster II® 9014 Safety Gloves, 60981 Series
Handschoenen
The Sharpsmaster II® 9014 safety glove utilises high-performance SuperFabric® material technology which is engineered to provide high levels of needle and cut protection.
  • High cut and needle resistance
  • SuperFabric® material protects against hazards
  • Wrinkle rubber palm coating over cotton blend knit
  • Seamless construction delivers high comfort and dexterity
  • Single-glove needle solution with incredible dexterity and comfort
Designed to offer exceptional needle and cut protection, this glove has been rigorously tested with 25-gauge needles, ensuring unparalleled safety for users in a single, high-performance solution. Three layers of SuperFabric® material protect against hazards through a configuration of tiny guard plates layered onto performance fabrics. Wrinkle rubber palm coating provides superior grip and abrasion resistance in wet conditions, and a knit wrist helps prevent dirt and debris from entering the glove. Seamless construction allows enhanced comfort while maintaining dexterity and tactility. The cotton blend shell is soft and dexterous.