Chemical Protection Gloves, u-fit ft 60166
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6016606 6016607 6016608 6016609 6016610
UVEX6016606EA 14.9 EUR
UVEX6016606 UVEX6016607 UVEX6016608 UVEX6016609 UVEX6016610
Chemical Protection Gloves, u-fit ft 60166
Handschoenen
The uvex u-fit ft is characterised by its excellent mechanical strength and also offers reliable protection against oil and dirt for light assembly activities.
  • Outstanding tactile feel
  • Very good fit
  • Extremely high flexibility
Thin and reliable NBR disposable safety gloves (0,10 mm).