Human ADAMTS13 ELISA Kits
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB234559-96 AB234559-1X384
ABCAAB234559-96EA 1000 EUR
ABCAAB234559-96 ABCAAB234559-1X384
Human ADAMTS13 ELISA Kits
Testen ELISAs
Human ADAMTS13 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human ADAMTS13 with a sensitivity of 16 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput. - Cited in over 10 citations.