Human VCAM-1 ELISA Set (without plates)
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB47355-480 AB47355-10X96 AB47355-15X96
ABCAAB47355-480EA 1140 EUR
ABCAAB47355-480 ABCAAB47355-10X96 ABCAAB47355-15X96
Human VCAM-1 ELISA Set (without plates)
Testen ELISAs
Human VCAM-1 ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human VCAM-1 ELISA Set (without plates) in Human in plasma, cell culture supernatant, serum samples.