TMB ELISA Substrate (High Sensitivity)
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB171523-100 AB171523-1000
ABCAAB171523-100EA 100 EUR
ABCAAB171523-100 ABCAAB171523-1000
TMB ELISA Substrate (High Sensitivity)
Testen ELISAs
Abcam's TMB ELISA Substrate (High Sensitive) detects horseradish peroxidase (HRP) activity and contains 3,3',5,5'-tetramethylbenzidine in a mildly acidic buffer.