Human CTGF ELISA kit
Leverancier: AbcamTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Human CTGF ELISA kit is a single-wash 90-min Simplestep used to quantify Human CTGF with a sensitivity of 64 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput.