Human CHIT1 ELISA Kit
Leverancier: AbcamTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Human CHIT1 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human CHIT1 with a sensitivity of 4,63 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput.