Human CHIT1 ELISA Kit
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB246541-1X96 AB246541-1X384
ABCAAB246541-1X96EA 944 EUR
ABCAAB246541-1X96 ABCAAB246541-1X384
Human CHIT1 ELISA Kit
Testen ELISAs
Human CHIT1 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human CHIT1 with a sensitivity of 4,63 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput.