Human IL-7 ELISA Kit
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB46031-96 AB46031-2X96
ABCAAB46031-96EA 969 EUR
ABCAAB46031-96 ABCAAB46031-2X96
Human IL-7 ELISA Kit
Testen ELISAs
Human IL-7 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-7 in Human in Plasma, Cell culture supernatant, Serum samples.