b.safe Y-Connection Tubes for Waste Pipes
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R1196-01 R1197-01
BOHLR1196-01EA 272 EUR
BOHLR1196-01 BOHLR1197-01
b.safe Y-Connection Tubes for Waste Pipes
Fittingen en connectoren
Y-shaped connector made of PE-EX.
  • Made of conductive PE-EX
  • Lateral connection with slope
  • Conductivity: 1000 kOhm
Lateral connections include a slope of 2° as well as union nut and clamp ring to insert and clamp b.safe waste tubes with O.D. 32 mm. Central connection either with male thread GL 25 to assemble b.safe waste tubes or straight tube O.D. 32 mm.