TriLink® Pseudouridine-5'-Triphosphate
Leverancier: TriLink BioTechnologies
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N-1019-10
N-1019-1
N-1019-5
SOLUN-1019-10EA
472
EUR
SOLUN-1019-10
SOLUN-1019-1
SOLUN-1019-5
TriLink® Pseudouridine-5'-Triphosphate
Reagentia voor nucleïnezuren
Nucleotiden
This nucleotide consists of a modified base, a ribose, and a 5′ triphosphate group.
- Can be incorporated in in vitro transcription for modification of synthesised RNA for desired characteristics
- ≥97% purity by AX-HPLC analysis, provided as 100 mM in H₂O
- Grade: RUO
Pseudouridine has been found to enhance base stacking and translation. Pseudouridine-5'-triphosphate (Pseudo-UTP) is used to impart desirable mRNA characteristics such as increased nuclease stability.
Opgelet: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.