TriLink® Guanosine-5'-Triphosphate
Leverancier: TriLink BioTechnologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N-1503-10 N-1503-100 N-1503-25
SOLUN-1503-10EA 25.3 EUR
SOLUN-1503-10 SOLUN-1503-100 SOLUN-1503-25
TriLink® Guanosine-5'-Triphosphate
Reagentia voor nucleïnezuren Nucleotiden
This nucleotide consists of an unmodified base, a ribose, and a 5′ triphosphate group for synthesis of RNA molecules.
  • Unmodified ribonucleotide for RNA synthesis
  • >99% purity by AX-HPLC analysis
  • Provided as 100 mM in H₂O, pH 7,5
  • Grade: RUO
Opgelet: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7154242f4b60e6c73b4f41f62dc17d76bb444609&vid=7154242f4b60e6c73b4f41f62dc17d76bb444609&prid=web&clid=SAPDAM