TriLink® 7-Deaza-2'-deoxyguanosine-5'-Triphosphate
Leverancier: TriLink BioTechnologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N-2011-1 N-2011-10 N-2011-5
SOLUN-2011-1EA 92.4 EUR
SOLUN-2011-1 SOLUN-2011-10 SOLUN-2011-5
TriLink® 7-Deaza-2'-deoxyguanosine-5'-Triphosphate
Reagentia voor nucleïnezuren Nucleotiden
This deoxyguanosine analog lacks the N7 nitrogen found in the natural guanine nucleobase. Replacement of this nitrogen atom with a carbon atom results in diminished structure of the major groove in DNA and may disrupt base stacking, but does not interfere with Watson-Crick base pairing. This is desirable for applications such as amplification and sequencing of GC-rich nucleic acid sequences.
  • Grade: RUO
Opgelet: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a8821bf0990e8e4a544423c92afacc35f079189&vid=6a8821bf0990e8e4a544423c92afacc35f079189&prid=web&clid=SAPDAM