TriLink® Biotin-16-Aminoallyl-2'-deoxyuridine-5'-Triphosphate
Leverancier: TriLink BioTechnologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N-5001-050 N-5001-1
SOLUN-5001-050EA 200 EUR
SOLUN-5001-050 SOLUN-5001-1
TriLink® Biotin-16-Aminoallyl-2'-deoxyuridine-5'-Triphosphate
Reagentia voor nucleïnezuren Nucleotiden
Biotin-16-Aminoallyl-2'-deoxyuridine can be enzymatically incorporated into DNA via PCR, nick translation, primer extension using Klenow(exo-) DNA polymerase, and terminal deoxynucleotidyl transferase (TdT). Biotinylated cDNA can be prepared with this NTP using Moloney Murine Leukemia Virus (M-MLV) reverse transcriptase.
  • Grade: RUO
Opgelet: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c825a111486248184c38eb32d00edf4801ce803&vid=5c825a111486248184c38eb32d00edf4801ce803&prid=web&clid=SAPDAM