TriLink® CleanAmp® 7-Deaza-dGTP
Leverancier: TriLink BioTechnologiesTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
CleanAmp® dNTPs are chemically-modified nucleotides designed to improve specificity, sensitivity, and robustness of your PCR.
- Improves PCR specificity, sensitivity, and robustness
- Provides an alternative approach to hot-start PCR without a need for a specially designed DNA polymerase
- Grade: RUO
The CleanAmp® 7-deaza-dGTP:
- Contains CleanAmp® 7-deaza deoxyguanosine 5’ triphosphate in a 2 µmol or 10 µmol format
- Is ideal for GC-rich PCR (~60% GC) due to promoting the template’s strand separation by lowering its melting temperature
- Enables hot-start PCR for specificity and sensitivity, with a thermolabile protecting group at the 3’ hydroxyl of the nucleotides (CleanAmp® dNTP benefit)
- Has been QC analysed by AX-HPLC, RP-HPLC, 31P NMR, mass spec, and a standardised PCR assay
Opgelet: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.