TriLink® 2',3'-Dideoxyguanosine-5'-O-(1-Thiotriphosphate)
Leverancier: TriLink BioTechnologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N-8011-1 N-8011-10 N-8011-5
SOLUN-8011-1EA 94.3 EUR
SOLUN-8011-1 SOLUN-8011-10 SOLUN-8011-5
TriLink® 2',3'-Dideoxyguanosine-5'-O-(1-Thiotriphosphate)
Reagentia voor nucleïnezuren Nucleotiden
2',3'-Dideoxyguanosine-5'-O-(1-Thiotriphosphate) is a sugar modified nucleoside triphosphate where the 2' and 3' hydroxyl groups are absent, resulting in chain termination. This ddGTP analog also contains a 1-Thiotriphosphate which results in a nuclease resistant phosphorothioate linkage. The inability of polymerases to extend from a dideoxy nucleotide causes chain termination and is useful in antiviral research and in a variety of biotechnology applications.
  • Grade: RUO
Opgelet: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18b99651dace6dfa44b7dd2c8d8292ea881f1e2a&vid=18b99651dace6dfa44b7dd2c8d8292ea881f1e2a&prid=web&clid=SAPDAM