TriLink® CleanCap® OVA mRNA
Leverancier: TriLink BioTechnologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L-7610-100 L-7610-1000 L-7610-5
SOLUL-7610-100EA 276 EUR
SOLUL-7610-100 SOLUL-7610-1000 SOLUL-7610-5
TriLink® CleanCap® OVA mRNA
Eiwitexpressie Reagentia voor eiwitexpressie
Ovalbumin (OVA) is a member of the serpin superfamily and the predominant glycoprotein found in egg whites. It is a commonly used antigen for immunisation and biochemical studies and is an established model allergen for airway hyper-responsiveness.
  • Capped with CleanCap® AG cap analog, no base modifications in the sequence
  • Cap 1 structure with >95% capping efficiency for superior in vivo activity compared to Cap-0 mRNAs
  • Manufactured with TriLink’s proprietary CleanScript® method to lower dsRNA for improved performance
  • Ready to use in downstream mRNA applications and formulations
  • Grade: RUO
This mRNA is capped using CleanCap® Reagent AG, TriLink's co-transcriptional capping technology, which results in the naturally occurring Cap-1 structure with >95% capping efficiency. It is manufactured using TriLink’s CleanScript® method, polyadenylated, and optimised for mammalian systems. It mimics a fully processed mature mRNA.
CleanCap Reagent AG produces a base-modified Cap-1 mRNA, which shows superior in vivo activity compared to Cap-0 mRNA produced by legacy capping methods such as mCap or anti-reverse cap analog (ARCA).
CleanScript method is TriLink’s proprietary in vitro transcription process that is optimised to minimise dsRNA formation and increase mRNA activity in vivo.
Opgelet: Item will be delivered at or below −40 °C and should be stored at or below −40 °C.