Reagent Reservoirs with Cover
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
701460 701462 701464
BRND701460EA 94.6 EUR
BRND701460 BRND701462 BRND701464
Reagent Reservoirs with Cover
Reagensreservoirs
For manual pipetting and automation. PP, 40 ml working volume, V-bottom, low dead volume, graduated.
  • CERTIFIED LIFE SCIENCE QUALITY: Virgin raw materials
  • Manufactured under controlled room conditions
  • Intensive in-process controls
  • Visual inspections
  • Batch management
  • Final product inspection
  • Proven functionality