BRAND® Pipetting Robot Liquid Handling Station
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
709400 709402
BRND709400EA 0 EUR
BRND709400 BRND709402
BRAND® Pipetting Robot Liquid Handling Station
Automated Liquid Handling Systems
The Liquid Handling Station provides maximum flexibility for small and medium sample throughput and varied pipetting tasks. It closes the gap between electronic pipettes and large, expensive equipment.
  • Extremely versatile, e.g. for PCR, enzyme tests and many other applications
  • Quick and uncomplicated method creation
  • Maximum performance at an attractive price
Seven ANSI / SLAS formatted workplaces can be freely assigned, including PCR tubes and strips, microcentrifuge tubes, reservoirs, and 96- or 384-well plates. Liquid ends are available in single-