VWR® Accessory Shelf Kits for Ultra-Low Temperature Freezers, Avantor
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SK700TSX SK500TSX SK600TSX
HERASK700TSXEA 351 EUR
HERASK700TSX HERASK500TSX HERASK600TSX
VWR® Accessory Shelf Kits for Ultra-Low Temperature Freezers, Avantor
Vriezers Ultra lage temperatuur vriezers
Organize your ULT freezer storage with additional interior stainless steel shelves.
  • Stainless steel shelf kit by freezer size
  • Includes shelf clips for installation
Certificaten:
cULus and CE.
Verpakking:
One ULT shelf and installation clips provided.