3-in-1 Flow-Through Cells for Fluorescence and Absorption Measurements
Leverancier: HELLMA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
176-761-85-40 176-762-15-40
HELL176-761-85-40EA 1640 EUR
HELL176-761-85-40 HELL176-762-15-40
3-in-1 Flow-Through Cells for Fluorescence and Absorption Measurements
Cuvetten Cuvetten voor spectrofotometers
Hellma® high precision 3-in-1 all-quartz flow-through cells for absorption and fluorescence measurements in the UV/Vis range, 5×2,5 mm and 3×1,5 mm path length, with 2 screw caps and FEP tubing.
  • Cell made of high performance quartz glass
  • Wavelength range from 200 to 2500 nm
  • Path length: 5×2,5 mm and 3×1,5 mm
  • Centre height: 8,5 or 15 mm
  • With 2 screw connections M6 and FEP tubing
  • Volume: 50 or 140 μl
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers.
Highest precision: Hellma® precision cells