Steridon Sterile Coveralls with Hood
Leverancier: ISOFIELD IRELAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
80111M 80111S 80111XS 80111XXS 80111L 80111XL 80111XXL 80111XXXL
ISOF80111MEA 489 EUR
ISOF80111M ISOF80111S ISOF80111XS ISOF80111XXS ISOF80111L ISOF80111XL ISOF80111XXL ISOF80111XXXL
Steridon Sterile Coveralls with Hood
Wegwerpkleding
The tri-laminate Steridon cleanroom coverall is aseptically folded and can be donned in 60 seconds or less.
  • Sterile cleanroom coverall with hood
  • Latex-free, enclosed and elasticated face, wrists, and ankles
  • Ergo-fit design at back and shoulders allowing greater range of motion
  • Aseptically presented fold
  • Colour-coded left and right inner tabs to facilitate donning
  • Removable external tabs at zipper to ensure sterile field is maintained
  • Validated for sterility by X-Ray
  • Sterility Assurance Level (SAL) 10⁻⁶
Steridon cleanroom coveralls are validated sterile via X-ray and tested against chemo to ASTM F 739. The anti-static, low-linting, viral barrier provides essential protection in Grade A/B environments for all cleanroom activities.
Verpakking:
Coveralls are packaged 1 piece per sealed inner bag, 1 inner bag per sealed outer bag, 20 outer bags per carton liner with twist tie, 20 pieces per carton (XXS to XXL); 15 pieces per carton (3XL to 5XL).