Steridon Sterile Coveralls with Integrated Facemask
Leverancier: ISOFIELD IRELAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
80231XS 80231XXS 80231L 80231M 80231S 80231XL 80231XXL 80231XXXL
ISOF80231XSEA 516 EUR
ISOF80231XS ISOF80231XXS ISOF80231L ISOF80231M ISOF80231S ISOF80231XL ISOF80231XXL ISOF80231XXXL
Steridon Sterile Coveralls with Integrated Facemask
Wegwerpkleding
The Isofield Steridon cleanroom coverall with integrated facemask is aseptically folded and can be donned in 60 seconds or less.
  • Sterile cleanroom coverall with integrated facemask
  • Latex-free, enclosed and elasticated face, wrists, and ankles
  • Ergo-fit design at back and shoulders allowing greater range of motion
  • Integrated facemask with a guided tie-on system to facilitate easy donning and secure fit
  • Aseptically presented fold
  • Colour-coded left and right inner tabs to facilitate donning
  • Removable external tabs at zipper to ensure sterile field is maintained
  • Validated for sterility by X-Ray
  • Sterility Assurance Level (SAL) 10⁻⁶
Steridon cleanroom coveralls are validated sterile via X-ray and tested against chemo to ASTM F 739. The anti-static, low-linting, viral barrier provides essential protection in Grade A/B environments for all cleanroom activities.
Verpakking:
Coveralls are packaged 1 piece per sealed inner bag, 1 inner bag per sealed outer bag, 20 outer bags per carton liner with twist tie, 20 pieces per carton (XX to XXL); 15 pieces per carton (3XL to 5XL).