Samplicity® G2 Filtration System and Filters
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SAMP2SYSB SAMP2SYSG SAMP2GVNB SAMP2HVNB SAMP2GNNB SAMP2HNNB SAMP2HPNB
MILFSAMP2SYSBEA 1850 EUR
MILFSAMP2SYSB MILFSAMP2SYSG MILFSAMP2GVNB MILFSAMP2HVNB MILFSAMP2GNNB MILFSAMP2HNNB MILFSAMP2HPNB
Samplicity® G2 Filtration System and Filters
Filters Filters zonder spuit
Vacuum filtration system for sample preparation for HPLC. Samplicity® G2 filtration system enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials.
  • Minimal hold-up volume
  • Filter and seal integrity
  • Low extractables, low binding
  • Integrated funnels for easiest sample loading
  • Filters in perforated strips of 4 for fastest setup
  • Lower ergonomic impact than manual syringe filtration
  • Non sterile
The adapter funnels provided for the Samplicity® G2 system are made of the same polypropylene as our Millex® filter housings. Together, the filter plus adapter are HPLC-certified to be low in extractable impurities.
Filtration for HPLC sample prep without syringes. Sample is added to an adapter funnel on a standard Millex® 33 mm filter (Millex® Filters for the Samplicity® G2 System) or an integrated funnel and filter (Millex Samplicity® filter).
Applications: HPLC; Filtration of viscous or particulate laden samples for HPLC. Analytical sample prep.
Polypropylene with polyester and silicone components. Components: Base, lid, vial tray, waste try and tubing.
Dimensions: 445L×102W×91H mm
Weight: 1,32 kg
Opgelet:
For general laboratory use only.