ESD Chairs, Texon
Leverancier: bimos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9173E-4000 9175E-4000
132-1469EA 698 EUR
132-1469 132-1470
ESD Chairs, Texon
Stoelen
In sensitive ESD work areas in the high-tech industry, first-class equipment is essential to provide reliable protection against electrostatic discharge. The ESD Texon fulfils the strict requirements of the European ESD standard EN 61340-5-1 and therefore offers maximum safety. The chair also makes a clear statement with its ergonomic design and high level of comfort.
  • Intuitive and simple operation
  • Seat height adjustment
  • Auto-Motion technology for active dynamic sitting and optimum support in any sitting position (Can be fixed in three settings)
  • The Flex-Back on the lower backrest increases the range of movement to the rear
  • Automatic weight adjustment
  • Extra thick seat upholstery for outstanding seating comfort
  • Replaceable upholstery system allows cushions to be easily removed and replaced as required
  • Aluminium base (ø 700 mm), anthracite
  • Max. load capacity 150 kg
Hygienic design - Smooth, closed surfaces and seamless design. Clean cover; Covers the mechanism for particularly easy cleaning. Disinfectable, resistant to chemicals and easy to keep clean.
Skintec is the innovative upholstery that combines first-class seating comfort with exceptional ease of cleaning and durability The surface is resistant to all disinfectants. Skintec is manufactured in a particularly sustainable and environmentally friendly way.
Certificaten:
European ESD standard EN 61340-5-1.