Cleanroom Chairs, Texon
Leverancier: bimos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9175R-4000 9175R-4002 9173R-4000 9173R-4002 9173R-4007 9175R-4007
132-1475EA 858 EUR
132-1475 132-1476 132-1472 132-1473 132-1474 132-1477
Cleanroom Chairs, Texon
Stoelen
Texon high-tech and cleanroom chair. Unique and equipped for everything - The Texon cleanroom is the chair for the high-tech industry. Cleanroom-qualified for air purity class 3 in accordance with DIN EN ISO 14644-1 and ESD-compliant in accordance with EN 61340-5-1, in hygienic design and with maximum ergonomic comfort - the Texon has everything that is important for demanding high-tech workstations.
  • Intuitive and simple operation
  • Seat height adjustment
  • Auto-Motion technology for active dynamic sitting and optimum support in any sitting position (Can be fixed in three settings)
  • The Flex-Back on the lower backrest increases the range of movement to the rear
  • Automatic weight adjustment
  • Extra thick seat upholstery for outstanding seating comfort
  • Replaceable upholstery system allows cushions to be easily removed and replaced as required
  • Aluminium base (ø 700 mm), silver grey
  • Max. load capacity 150 kg
Hygienic design - Smooth, closed surfaces and seamless design. Clean cover; Covers the mechanism for particularly easy cleaning. Disinfectable, resistant to chemicals and easy to keep clean.
Skintec is the innovative upholstery that combines first-class seating comfort with exceptional ease of cleaning and durability The surface is resistant to all disinfectants. Skintec is manufactured in a particularly sustainable and environmentally friendly way.
Certificaten:
Cleanroom-qualified for air purity class 3 in accordance with DIN EN ISO 14644-1 and ESD-compliant in accordance with EN 61340-5-1. GMP-compliant.