Tork Reflex™ M3 Mini Centrefeed Dispensers
Leverancier: Essity
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
473167 473177
SCAA473167EA 56.2 EUR
SCAA473167 SCAA473177
Tork Reflex™ M3 Mini Centrefeed Dispensers
Dispensers voor doekjes
Suitable for tough environments and small spaces, this centrefeed dispenser is robust yet compact. Boost hygiene levels in a workspace with a design that's easy to clean.
  • Reduce cross-contamination by touching only what is taken
  • Avoid the hassle of running out of paper with the handy run out indicator
  • Design: Performance
Busy work areas need durable products - Tork Reflex™ Centrefeed Dispenser is robust enough for heavy daily use and provides a solution for both hand and surface wiping. Its compact design features a rotating nozzle that suits professional spaces of all sizes. Waste is reduced, thanks to single-sheet dispensing which controls consumption by up to 37% compared with the Tork® Centrefeed system. Run out indicator further improves efficiency, enabling refills to be planned in advance. Carbon neutral - produced with certified renewable electricity and compensated with climate projects.