Tork W7 Heavy-Duty Cleaning Cloths
Leverancier: Essity
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
530172 530272
SCAA530172EA 71.7 EUR
SCAA530172 SCAA530272
Tork W7 Heavy-Duty Cleaning Cloths
Schoonmaakdoekjes Herbruikbare reinigingsdoeken Doeken voor delicate taken
Optimise consumption of the disposable cleaning cloths with the one-at-a-time dispensing feature. The Handy Box's large opening means cloths are easy to access, even when wearing gloves.
  • Cut time and waste and ensure staff have as many cleaning cloths as they need close by, thanks to the portable handy box
  • Improve brand's sustainability - packaging is made out of 100% recycled fibres
Get tough on dirt and make hard-to-reach spaces sparkle with the Tork heavy-duty cleaning cloth. This thick disposable cleaning cloth is strong and absorbent enough to tackle liquids, oil and grease. Use with most solvents for added ease, while helping the environment and staff health with the exelCLEAN® material - it uses up to 41% less solvent than rags. Plus, we've lowered the CO₂ emissions for Tork exelCLEAN® by 28% since 2011. Workflow efficiency is improved and time saved with the lightweight and portable Tork handy box. Cut waste, too, with the one-at-a-time dispensing. (Life Cycle Analysis) LCA conducted by Essity and IVL Svenska Miljöinstitutet in April 2021
Certificaten: FSC MIX Credit, SA-COC-008266.