Endothelial Basal Medium, MCDB 131
Leverancier: US Biological
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E3000-01B-100L E3000-01B-10L E3000-01B-25L E3000-01B-50L
USBIE3000-01B-100LEA 0 EUR
USBIE3000-01B-100L USBIE3000-01B-10L USBIE3000-01B-25L USBIE3000-01B-50L
Endothelial Basal Medium, MCDB 131
Celcultuurmedia
MCDB 131 is a reduced serum-supplemented medium for the culture of human microvascular endothelial cells. This formulation has been reported to support clonal growth of human omental microvascular cells when supplemented with 0,7% dialysed FBS, 10 ng/ml EGF, and 1 μg/ml hydrocortisone.
  • Pale orange to orange, homogeneous, free-flowing powder
  • Pink to red or orange, clear, complete solubility
  • Endotoxin ≤1 EU/ml
Store powdered media at RT. Opened bottles should be capped tightly and kept in a dark, low humidity environment. Prepared media should be kept refrigerated and used within a short period of time. Shipping temperature RT.