Mechanical Gloves for Work in Dry Environments
Leverancier: DELTA PLUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VE702P/8 VE702P/10
DELPVE702P/8EA 2.8 EUR
DELPVE702P/8 DELPVE702P/10
Mechanical Gloves for Work in Dry Environments
Handschoenen
Mechanical gloves ideal for work in dry environments. The anti-slip properties of the polyurethane coating provide good grip.
  • Knitted coated gloves
  • Support: Tricot - polyester
  • Benefits element: Reach
  •  
  •  
Applications: Public Works, finishing works/craftsmanship, maintenance, automotive, temporary work, water and waste treatment, power energy, storage.
Certificaten:
EN 388:2016+A1:2018.