Top-Rack Tripods
Leverancier: NEOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2-2566 2-2563 2-2562 2-2561
NEOL2-2566EA 6.52 EUR
NEOL2-2566 NEOL2-2563 NEOL2-2562 NEOL2-2561
Top-Rack Tripods
Rekken Tube Racks
Colour top-rack tripods, 80 seats
  • Made of polypropylene, with two holders
  • 80-seater
  • Designed for tubes with a capacity of 1,5 and 2 ml
Resistant to temperatures from −80 to +125 °C. Can be autoclaved. Tripods can be stored in properly prepared boxes with a lid (one box for one tripod)