Combi-Rack for Test Tubes
Leverancier: NEOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2-2573 2-2575 2-2572
NEOL2-2573EA 30.4 EUR
NEOL2-2573 NEOL2-2575 NEOL2-2572
Combi-Rack for Test Tubes
Rekken Tube Racks
Combi rack for test tubes, 96-position.
  • Double-sided test tube rack with lid
  • Available in 7 colours
  • Made of PP
  • Autoclavable up to 121 °C
On the upper side there are holes for test tubes with a capacity of 1,5 and 2 ml, on the lower one places for test tubes with a capacity of 1,5 and 2 ml. 0,5 ml.