Stainless Steel Heavy Duty Tables with Perforated Top
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TLWB-RR/P18X90 TLWB-RR/P12X60
TEKMTLWB-RR/P18X90EA 0 EUR
TEKMTLWB-RR/P18X90 TEKMTLWB-RR/P12X60
Stainless Steel Heavy Duty Tables with Perforated Top
Meubilair Tafels
The stainless steel rear tie bar heavy duty table with perforated top is an ideal surface for industries that require sterile, hygienic and contamination resistant surfaces. It is more sturdy with a heavier payload for items that are of a heavier weight.
  • Frame in 304 L-grade stainless steel
  • Brushed dull polish finish
  • 304-grade perforated top with a silicone seal
  • 304-grade perforated undershelf
  • 4× stainless steel feet with non-marking floor levelling rubber base
  • Payload of up to 250 kg
  • Floor clearance to undershelf: 155 mm
With both the top and optional shelf perforated for optimum airflow and an all 304 L-grade stainless steel construction, it's the perfect hygienic work surface for any clean environment.