HPTLC Software visionCATS
Leverancier: CAMAG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
028.0000 028.2000
CAMA028.0000EA 3640 EUR
CAMA028.0000 CAMA028.2000
HPTLC Software visionCATS
Software Laboratoriumsoftware
VisionCATS is built for simplicity and intuitive operation, streamlining the entire HPTLC workflow by controlling both HPTLC PRO Modules and HPTLC instruments while efficiently managing data.
  • Comparison viewer
  • Image enhancement tools
  • Scanning densitometry and spectral evaluation
  • Quantitative analysis
  • HPTLC method library
  • Regulatory compliance
With its user-friendly interface, visionCATS guides users smoothly through the entire chromatographic process, from sample setup to report generation. Starting is easy - select a default method, fill in the sample table, choose a developing solvent, and select a derivatisation reagent. Detection parameters can be customised as needed, and visionCATS leads users through each step.
Creating custom methods is equally straightforward. Users can easily select the required steps, and the sample-oriented design enables the creation of virtual plates by combining tracks from different plates. This feature is especially useful for batch-to-batch comparisons and long-term stability testing.
Finding relevant samples is made faster with a robust search tool in the file explorer. Users can search by text, date, samples, methods, or analysis files, with extended preview options. Custom search filters can also be created, allowing for even more streamlined navigation and data retrieval.
CAMAG® HPTLC Software visioncats: Basic version: Including access and control of all instruments - 1 server, 1 client Instrument diagnostics (xQ), analytical reports - access to method library. Needs to be purchased separately and is not included in any ultimate package.
CAMAG® HPTLC Software visionCATS: Visualiser ultimate package: Combining all CAMAG visualiser packages, including: Visualiser qualitative package (028.2010), Visualiser enhanced evaluation package (028.2020); CAMAG HPTLC Software visionCATS Basic version (028.0000) needs to be purchased separately.
Key benefits:
Faster, more accurate identification
Efficient data interpretation
Reliable detection of adulteration and contaminants
Accelerated research and development