gigasept® AF Forte
Leverancier: SCHUELKE MAYR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70003412 70003413
SCUE70003412EA 0 EUR
SCUE70003412 SCUE70003413
gigasept® AF Forte
Reinigingsmiddelen Desinfectiemiddelen
gigasept® AF forte is an aldehyde-free preparation for cleaning and disinfection of rigid and flexible endoscopes as well as surgical instruments.
  • Aldehyde-free
  • Suitable for flexible and rigid endoscopes
  • Shortened action time in ultrasonic baths
  • Patented substance combination
Cleaning and disinfection of flexible and rigid endoscopes, surgical instruments, anaesthesia equipment and breathing masks. Green colour, liquid form, pH 9,8 to 10.
New Product
Order Now
Specificaties
Environmentally Preferable