Stericlin® Non-Woven Wrapping Material
Leverancier: VP Medical Packaging
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3FVLI330104 3FVLI330106 3FVLI330108 3FVLI330114 3FVLI330116 3FVLI330124 3FVLI330126 3FVLI330128 3FVLI330118 3FVLI330112 3FVLI330120
129-1370EA 128 EUR
129-1370 129-1371 129-1372 129-1374 129-1375 129-1378 129-1379 129-1380 VEPA3FVLI330118 VEPA3FVLI330112 129-1377
Stericlin® Non-Woven Wrapping Material
Sterilisatieverpakkingen
Wrapping material non-woven from stericlin® The combination of cellulose and synthetic fibres.
Stericlin® wrapping material made of non-woven is known for its very good draping properties together with high mechanical stability. They meet the requirements of DIN EN ISO 11607 and DIN EN 868-2 and are suitable as inner wrapping for sterile containers and for packaging medical devices such as instrument trays or linen.
Stericlin® non-woven consists of cellulose and synthetic fibres, whereby the cellulose fibres promote drying and the synthetic fibres provide strength and softness. As a sterile barrier system, it prevents contamination from seeping through and is thus ideally suited for aseptic presentation.
Certificaten:
DIN EN ISO 11607 and DIN EN 868-2.