BradyPrinter i5300 Industrial Label Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
151274 151275 151283 151284 322900 322891 322894 322897 322899 322890 322893
BRDY151274EA 1645 EUR
BRDY151274 BRDY151275 BRDY151283 BRDY151284 BRDY322900 BRDY322891 BRDY322894 BRDY322897 BRDY322899 BRDY322890 BRDY322893
BradyPrinter i5300 Industrial Label Printers
Printers Label Printers
Zero calibration, zero wasted labels, with higher-volume duty cycle and ultra-precise printing. Prints on a very wide range of B30/B33 materials with exceptional accuracy on labels as small as 5 mm. For ultimate high-mix printing with 20 second no-adjustment material changeovers, built-in auto-cutting and optional save-to-printer job lists for a quick way to print commonly used labels and overwrite-protected labels away from the PC. When focused on efficiency, every process counts - let simplicity drive productivity.
  • No wasted labels - prints on the first one
  • i5300 - 20-second supply changeovers
  • 300 and 600 dpi printer solution models
  • One click labels part recognition and setup
  • Faster print speeds for all job sizes
  • Rugged construction for mid to high volumes
  • Colour: Blue, grey
Taking just seconds to set up and load, it offers a faster, easier start-to-print process. it’s designed for efficiency - no setup, no adjustments and no waste. Time-saving features like a print queue and interrupt function. Plus, get precision printing on 1300 parts. All this can reach high-mix, high-volume printing goals.
Printer features:
Solid construction with metal framing and gearing for higher throughput and workhorse durability
Built-in, programmable auto-cutter
Big 7" colour touchscreen
Intuitive interface
Smart media rolls
6 to 10 mm label widths
Easy loading; 20-second changeovers
Double-side wire sleeve printing
Fast print speed, connectivity and ports
Field-replaceable print roller and printhead
Print queue
No calibration is required, so there are no wasted labels and no trial and error during setup. There are no sensor, heat or speed adjustments. Just drop, lock and print to reduce setup time. Prints small fonts with excellent precision and accuracy for electronics, product and laboratory identification.
Software, printer and materials communicate to allow 1-click auto setup of labels. Get efficient, large-job printing with better performance and higher throughput. Metal-frame construction and internal print engine are designed for high performance.
Leveringsinformatie:
This i5300 Industrial label printer comes in a cardboard box. It ships with Brady WorkStation basic design suite, power cable, USB cable, stylus, cleaning kit, QuickStart guide, jump drive with how to videos and 3rd party win driver.