Cryogenic Labels for Inkjet Printers
Leverancier: GA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A4CIJSA-8WH A4CIJSA-12WH A4CIJSA-32WH A4CIJSA-23WH
GAINA4CIJSA-8WHEA 82.2 EUR
GAINA4CIJSA-8WH GAINA4CIJSA-12WH GAINA4CIJSA-32WH GAINA4CIJSA-23WH
Cryogenic Labels for Inkjet Printers
Etiketten
Inkjet printable cryogenic labels are ideal for identifying cryo vials, tubes, plastic bags, as well as cardboard and polycarbonate freezer boxes.
  • Temperature range: −196 to +121 °C (−321 to +250 °F)
  • Print eye-catching color logos, graphics, images, and data
  • Resists wiping with ethanol and surface sanitizing wipes
  • Require zero cure time (can be immersed in liquid nitrogen directly following application)
  • Resists extreme cryogenic and deep-freeze temperatures (−196 °C in a liquid phase and vapor phase Liquid Nitrogen and −80 °C)
These labels are designated for cryo storage in liquid nitrogen (−196 °C/−321 °F) and lab freezers (−80 °C/−112 °F) with eye-catching color logos, graphics, images, and data. Moreover, these durable labels also resist dry heat and standard steam autoclave sterilization cycles, and are perfect for a variety of uses in scientific laboratories.
The obtained printout offers an impressive resistance to harsh chemicals, such as alcohol, acetone, and xylene, as well as to wiping and rubbing with alcohol sanitizing wipes.
Sheet format labels can be printed with most standard desktop inkjet printers. They can also be written on with ballpoint pens, cryogenic markers, and ink stamped without smudging.