FARRAR® ULC Ultra-Low Chambers
Leverancier: TRANE TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ULC259-BWC50HZ ULC328-SAC50HZ ULC190-SAC50HZ ULC190-SWC50HZ ULC259-DAC50HZ ULC259-DWC50HZ ULC259-SWC50HZ ULC259-BAC50HZ ULC259-SAC50HZ ULC328-SWC50HZ
FARRULC259-BWC50HZEA 0 EUR
FARRULC259-BWC50HZ FARRULC328-SAC50HZ FARRULC190-SAC50HZ FARRULC190-SWC50HZ FARRULC259-DAC50HZ FARRULC259-DWC50HZ FARRULC259-SWC50HZ FARRULC259-BAC50HZ FARRULC259-SAC50HZ FARRULC328-SWC50HZ
FARRAR® ULC Ultra-Low Chambers
Vriezers Ultra lage temperatuur vriezers
Purpose-built for life science applications, FARRAR®’s ULC series including the Ultra-Low Chamber (ULC-190), Ultra-Low Chamber (ULC-328), and the new Ultra-Low Chamber (ULC-259) are the only −20 to −80 °C, forced-air, ultra-low-temperature freezers on the market with air and water-cooled options.
  • Optimized footprint - 50% less floorspace versus standard ULT configurations with six times the capacity (one ULC equals six ULTs)
  • Industry's fastest temperature recovery - 10× faster temperature recovery from door opening helps ensure product integrity
  • Integrated, fully redundant systems (control, refrigerations, airflow) - Eliminates need for backup freezer
  • Automatic defrost - Never worry about taking units out of service to manually defrost
  • Extremely tight temperature uniformity at any setpoint - At +4, −80 °C or any temperature in between, count on better than ±3 °C anywhere inside the ULC, even during a defrost cycle
  • Engineered with material handling in mind - Optional racking, shelving, drawers and carts optimise process and workflow (available for all models)
  • Equipment performance/temperature monitoring (MODbus) - Enables easy connection to nearly every building management and automation system (BAS/BMS)
Forced air convection: An industry game changer - FARRAR® pioneered forced air convection technology in its reach-in chambers. This key differentiator affects uniformity. The chambers uniformly force air, at the targeted temperature, throughout the chamber. This means FARRAR® freezer and freeze/thaw equipment is up to 10 times more efficient at removing heat than the traditional cold-wall heat displacement process. Recovery from door opening is critical to helping ensure product integrity. FARRAR® ULC Series chambers dual refrigeration units recover to −80 °C in less than 25 minutes from a 1 minute door opening, versus several hours experienced with competitor solutions.
Informatie over toebehoren:
Components included in the standard set-ups for each unit are listed below (with supplier part numbers). Please speak to your sales representative about other options and accessories available.
ULC-190 Standard Set-up, Shelves
* ULC 190 Chamber - #6700617
* Refrigeration System - #6700030 (air-cooled), #6700031 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* Standard ULC-190 Shelf Kit - #6700645
ULC-259 Standard Set-up, Shelves
* ULC 259 Chamber - #6700619
* Refrigeration System - #6700030 (air-cooled), #600031 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* ULC-259 Standard Shelf System - #6700775
ULC-259 Standard Set-up, Drawers
* ULC 259 Chamber - #6700619
* Refrigeration System - #6700030 (air-cooled), #600031 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* ULC-259 Drawer System - #6700761
ULC-259 Standard Set-up, Biobank
* ULC 259 Chamber - #6700619
* Refrigeration System - #6700030 (air-cooled), #600031 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* ULC-259 Biorack Assembly - #6700769
ULC-328 Standard Set-up, Pallet Rack
* ULC 328 Chamber - #6700615
* Refrigeration System - #6700030 (air-cooled), #600031 (water-cooled)
* Middle Box (Non-Monitoring) - #6000442
* ULC-328 Pallet Rack Assembly - #6700401