Metal Cases for Small Parts
Leverancier: hünersdorff GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
620700 620800
HUEN620700EA 0 EUR
HUEN620700 HUEN620800
Metal Cases for Small Parts
Zakken Equipment Bags and Instrument Cases
Small parts case made of powder-coated steel for individual fitting with PS insert boxes, dark blue.
  • Stable lever-action locks, lockable and lead sealable
  • Tightly closing lid avoids mixing of the contents
Lid lock is available against unintentional opening of the case.  Empty case for individual fitting with insert boxes (insert box height 63 mm).