Spiral Condensers
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.772.250 1.770.250.S
WITG1.772.250EA 219 EUR
WITG1.772.250 WITG1.770.250.S
Spiral Condensers
Condensors
Spiral Condensers with water connection 2× GL 14, made of borosilicate glass.