Brady® M510 Printer Kit with Vial Labels
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
324273 324274
BRDY324273EA 0 EUR
BRDY324273 BRDY324274
Brady® M510 Printer Kit with Vial Labels
Printers Label Printers
Create large 38,10 mm labels with edge-to-edge printing and a wide range of text sizes.
  • Print up to 38,10 mm per second to get work done faster
  • Backlit full-colour LCD graphics display for easier labelling in low-lit areas
  • Durably designed to withstand 1,2 m drops
  • The rechargeable NiMH battery prints up to 1700 labels on a single charge
With 250+ hassle-free drop-in cartridges and 26+ high-performance materials. Enter data with more accuracy as you work between an intuitive QWERTY keypad, a separate numeric keypad or a PC.
Ships with a licence for the brady workstation suite of your preference: product and wire identification, safety and facility identification, or lab identification, enabling easy creation of a wide range of labels and signs.
Applications include asset tracking, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, facility identification, general identification, inventory and inspection labelling, laboratory labelling, lean and 5s labelling, safety identification, warehouse marking, wire and cable labelling, barcode labelling, healthcare and hospital labelling.
Keyboard type: QWERTY
Certificaten:
UKCA; FCC.
Informatie over toebehoren:
M4-; M4C-; M5-; M5C-.
Bestelinformatie:
Two-year warranty.
Leveringsinformatie:
This M510 Printer ships in a cardboard box with a rechargeable NiMH battery pack, an AC adapter, B-595 vinyl label cartridge, B-499 nylon cloth label cartridge, a quick start guide, a USB cable, a licence for Brady Workstation Software suites and a USB flash drive with driver and user manual.