Blotting membranes, Amersham™ Hybond™ 0,45 PVDF
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
10600023 10600086 10600087 10600029 10600038 10600069 10600100 10600121
10600023.EA 419 EUR
10600023. 10600086. 10600087. 10600029. 10600038. 10600069. 10600100. 10600121.
Blotting membranes, Amersham™ Hybond™ 0,45 PVDF
Transfermembranen
Amersham™ Hybond™ P 0,45 PVDF is a 0,45 μm pore size hydrophobic membrane, with high physical strength, highly suitable for stripping and reprobing. For use in chemiluminescent and fluorescent detection methods for proteins of >Mr 20000.
  • Protein binding capacity over 200 μg/cm² for easy signal detection
  • High protein retention, even after harsh washing steps
  • Maximum capture of proteins during transfers minimises sample loss
  • Available in popular pre-cut sizes
For convenience, Western blotting sandwiches (10 sandwiches/package) are available. EachMore Product Information