Thrombin (biotinylated)

Leverancier: Merck Millipore (Novagen)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

69672-1000UN 69672-500UN
69672-1000UNEA 4340 EUR
69672-1000UN 69672-500UN
Thrombin (biotinylated)
Enzymen

Biotinylated Thrombin has covalently attached biotin for easy removal of the enzyme from cleavage reactions using immobilised streptavidin. This preparation is tested for activity using the same assay as for unmodified thrombin, and for >99% binding to streptavidin agarose.


Storage temperature: −20 °C.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR