Tris(hydroxypropyl)phosphine solution 0.5 M, Millipore®

Leverancier: Merck Millipore (Novagen)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: THPP, THP, Tris(3-hydroxypropyl)phosphine

71194-3 71194-4
71194-3EA 61.9 EUR
71194-3 71194-4
Tris(hydroxypropyl)phosphine solution 0.5 M, Millipore®
Tris(hydroxypropyl)phosphine

THP [Tris(hydroxypropyl)phosphine; C9H21O3P] is a water-soluble, odorless, sulfhydryl reducing agent that is more stable and effective than dithiothreitol (DTT).


Against oxidised glutathione, the reducing capacity of THP (0,232 meq/g) is greater than that of TCEP (0,182 meq/g). THP is also more resistant to air oxidation than DTT and is compatible with immobilized metal affinity chromatography with His•Bind® and Ni-NTA His•Bind resins at concentrations up to 1,0 mM. The common working concentration of THP for many applications is 1–5 mM. THP is provided as a ready-to-use 0,5 M solution in water.

Formule: C₉H₂₁O₃P
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00269835
CAS nummer: 4706-17-6
UN: 2922
ADR: 8 (6.1),II

Order Now

Specification Test Results

Purity >80%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR