Yeast extract, Veggie™
Leverancier: Merck Millipore (Novagen)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
71279-3
71279-3EA 227 EUR
71279-3
Yeast extract, Veggie™
Media voor microbiologie Additieven en supplementen voor microbiologie
Veggie yeast extract is a certified animal-free media component.
  • Ensures proper growth and maintenance of bacterial cells
  • Ideal for applications requiring minimal use of animal-derived materials
Used as a direct replacement for traditional yeast extract in bacterial growth media.